Learn More
Medchemexpress LLC Imatinib | 152459-95-5 | MFCD00868202 | C29H31N7O | 500 MG
SDP
Catalog No. 5000376450
Encompass_Preferred
Imatinib (STI571) is an orally bioavailable tyrosine kinases inhibitor that selectively inhibits BCR/ABL, v-Abl, PDGFR, and c-kit kinase activity. It functions by binding near the ATP binding site, which locks the kinase in a closed or self-inhibited conformation, thereby semicompetitively inhibiting the enzyme activity of the protein. Additionally, Imatinib is an inhibitor of SARS-CoV and MERS-CoV.
  • Selectively inhibits BCR/ABL, v-Abl, PDGFR, and c-kit kinase activity.
  • Functions by binding near the ATP binding site.
  • Inhibits enzyme activity of the protein semicompetitively.
  • Inhibitor of SARS-CoV and MERS-CoV.
  • For research use only.
Catalog No. 50-003-76450 Supplier Medchemexpress LLC Supplier No. HY15463500MG
Imatinib (STI571) is an orally bioavailable tyrosine kinases inhibitor that selectively inhibits BCR/ABL, v-Abl, PDGFR, and c-kit kinase activity. It functions by binding near the ATP binding site, which locks the kinase in a closed or self-inhibited conformation, thereby semicompetitively inhibiting the enzyme activity of the protein. Additionally, Imatinib is an inhibitor of SARS-CoV and MERS-CoV.
  • Selectively inhibits BCR/ABL, v-Abl, PDGFR, and c-kit kinase activity.
  • Functions by binding near the ATP binding site.
  • Inhibits enzyme activity of the protein semicompetitively.
  • Inhibitor of SARS-CoV and MERS-CoV.
  • For research use only.