Learn More
Medchemexpress LLC Imatinib | 152459-95-5 | MFCD05662257 | C29H31N7O | 10 G
SDP
Catalog No. 5000376208
Encompass_Preferred
Imatinib is an orally bioavailable tyrosine kinase inhibitor that selectively inhibits BCR/ABL, v-Abl, PDGFR, and c-kit kinase activity. It functions by binding near the ATP binding site, locking it in a closed or self-inhibited conformation, thereby semicompetitively inhibiting the enzyme activity of the protein. It is also an inhibitor of SARS-CoV and MERS-CoV and acts as an antineoplastic agent.
  • Orally bioavailable tyrosine kinase inhibitor
  • Selectively inhibits BCR/ABL, v-Abl, PDGFR, and c-kit kinase activity
  • Functions by binding close to the ATP binding site
  • Inhibits SARS-CoV and MERS-CoV
  • Acts as an antineoplastic agent
  • Used for the treatment of chronic myelogenous leukemia and gastrointestinal stromal tumours
Catalog No. 50-003-76208 Supplier Medchemexpress LLC Supplier No. HY1546310G
Imatinib is an orally bioavailable tyrosine kinase inhibitor that selectively inhibits BCR/ABL, v-Abl, PDGFR, and c-kit kinase activity. It functions by binding near the ATP binding site, locking it in a closed or self-inhibited conformation, thereby semicompetitively inhibiting the enzyme activity of the protein. It is also an inhibitor of SARS-CoV and MERS-CoV and acts as an antineoplastic agent.
  • Orally bioavailable tyrosine kinase inhibitor
  • Selectively inhibits BCR/ABL, v-Abl, PDGFR, and c-kit kinase activity
  • Functions by binding close to the ATP binding site
  • Inhibits SARS-CoV and MERS-CoV
  • Acts as an antineoplastic agent
  • Used for the treatment of chronic myelogenous leukemia and gastrointestinal stromal tumours