Learn More
Medchemexpress LLC HY-15463 5g , Imatinib STI571 CAS:152459-95-5 Purity:98%
SDP
Catalog No. 501492179
Encompass_Preferred
Medchemexpress, HY-15463 5g Imatinib STI571 CAS:152459-95-5 Formula:C29H31N7O IC50: ~100 nM (c-Kit, Bcr-Abl, and PDGFR) Purity:98% Imatinib is a known inhibitor of the c-Kit , Bcr-Abl , and PDGFR tyrosine kinases, inhibits the SLF-dependent activation of c-Kit wt kinase with IC 50 of ~100 nM, which is similar to the concentration requires for inhibition of Bcr-Abl and PDGFR. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2179 Supplier Medchemexpress LLC Supplier No. HY154635G
Medchemexpress, HY-15463 5g Imatinib STI571 CAS:152459-95-5 Formula:C29H31N7O IC50: ~100 nM (c-Kit, Bcr-Abl, and PDGFR) Purity:98% Imatinib is a known inhibitor of the c-Kit , Bcr-Abl , and PDGFR tyrosine kinases, inhibits the SLF-dependent activation of c-Kit wt kinase with IC 50 of ~100 nM, which is similar to the concentration requires for inhibition of Bcr-Abl and PDGFR. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.