Learn More
Medchemexpress LLC Imatinib acid | 2741007-77-0 | 99.7% | 551.64 g/mol | C31H33N7O3 | 5 MG
SDP
Catalog No. 5000221865
Encompass_Preferred
Imatinib acid is a research-grade small-molecule analogue of imatinib (CAS 2741007-77-0) supplied as a solid for laboratory use. It is intended for in vitro pharmacology, analytical reference, and chemical intermediate applications and is not for human or clinical use. Product documentation includes a data sheet and a safety data sheet for handling and storage guidance.
  • High purity (99.73%) suitable for analytical and research use.
  • Molecular formula C31H33N7O3 and molecular weight 551.64 g/mol.
  • Available in small packaged quantities for precise dosing.
  • Provided with a safety data sheet and product data sheet.
  • Suitable as a reference standard, metabolite, or intermediate in research workflows.
Catalog No. 50-002-21865 Supplier Medchemexpress LLC Supplier No. HY1356385MG
Imatinib acid is a research-grade small-molecule analogue of imatinib (CAS 2741007-77-0) supplied as a solid for laboratory use. It is intended for in vitro pharmacology, analytical reference, and chemical intermediate applications and is not for human or clinical use. Product documentation includes a data sheet and a safety data sheet for handling and storage guidance.
  • High purity (99.73%) suitable for analytical and research use.
  • Molecular formula C31H33N7O3 and molecular weight 551.64 g/mol.
  • Available in small packaged quantities for precise dosing.
  • Provided with a safety data sheet and product data sheet.
  • Suitable as a reference standard, metabolite, or intermediate in research workflows.