Learn More
Medchemexpress LLC Imatinib-d4 | 1134803-16-9 | 99.9% | 497.63 | C29H27D4N7O | 10 MG
SDP
Catalog No. 5000285951
Encompass_Preferred
Imatinib-d4 is a deuterium-labeled analogue of imatinib provided for research use as a stable isotope internal standard. It is intended for bioanalytical and pharmacokinetic applications where mass-shifted quantification improves accuracy and precision in LC-MS assays and metabolic studies.
  • Stable isotope labeled internal standard for LC-MS assays.
  • Four deuterium atoms create a clear mass shift for quantification.
  • High chemical purity for consistent analytical performance.
  • Suitable for pharmacokinetic, bioanalytical, and metabolic studies.
  • Available in small milligram quantities for laboratory use.
Catalog No. 50-002-85951 Supplier Medchemexpress LLC Supplier No. HY15463S110MG
Imatinib-d4 is a deuterium-labeled analogue of imatinib provided for research use as a stable isotope internal standard. It is intended for bioanalytical and pharmacokinetic applications where mass-shifted quantification improves accuracy and precision in LC-MS assays and metabolic studies.
  • Stable isotope labeled internal standard for LC-MS assays.
  • Four deuterium atoms create a clear mass shift for quantification.
  • High chemical purity for consistent analytical performance.
  • Suitable for pharmacokinetic, bioanalytical, and metabolic studies.
  • Available in small milligram quantities for laboratory use.