Learn More
Medchemexpress LLC Imatinib-d8 | 1092942-82-9 | 99.9% | 501.65 g/mol | C29H23D8N7O | 10 MG
SDP
Catalog No. 5000287429
Encompass_Preferred
Imatinib-d8 is a deuterium-labeled analogue of imatinib designed for use as an isotopic reference standard in analytical and pharmacokinetic studies. It retains the core structure of imatinib with eight deuterium atoms and is intended for use in mass spectrometry, LC-MS/MS assays, and ADME/PK research.
  • Deuterium-labeled analogue for isotopic internal standard applications.
  • High purity suitable for analytical and quantitative studies.
  • Optimized for LC-MS/MS and pharmacokinetic analyses.
  • Chemical formula C29H23D8N7O and molecular weight 501.65 g/mol aid in accurate mass calculations.
  • Provided as a research-grade reference material for assay development and validation.
Catalog No. 50-002-87429 Supplier Medchemexpress LLC Supplier No. HY15463S10MG
Imatinib-d8 is a deuterium-labeled analogue of imatinib designed for use as an isotopic reference standard in analytical and pharmacokinetic studies. It retains the core structure of imatinib with eight deuterium atoms and is intended for use in mass spectrometry, LC-MS/MS assays, and ADME/PK research.
  • Deuterium-labeled analogue for isotopic internal standard applications.
  • High purity suitable for analytical and quantitative studies.
  • Optimized for LC-MS/MS and pharmacokinetic analyses.
  • Chemical formula C29H23D8N7O and molecular weight 501.65 g/mol aid in accurate mass calculations.
  • Provided as a research-grade reference material for assay development and validation.