Learn More
Medchemexpress LLC HY-50946 1g , Imatinib (Mesylate) CAS:220127-57-1 Purity:>98%
SDP
Catalog No. 502259395
Encompass_Preferred
HY-50946 1g Imatinib (Mesylate) CAS: 220127-57-1 Imatinib Mesylate (STI571 Mesylate) is a tyrosine kinases inhibitor that inhibits c-Kit, Bcr-Abl, and PDGFR (IC50=100 nM) tyrosine kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9395 Supplier Medchemexpress LLC Supplier No. HY509461G
HY-50946 1g Imatinib (Mesylate) CAS: 220127-57-1 Imatinib Mesylate (STI571 Mesylate) is a tyrosine kinases inhibitor that inhibits c-Kit, Bcr-Abl, and PDGFR (IC50=100 nM) tyrosine kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.