Learn More
Medchemexpress LLC Imatinib (Mesylate) | 220127-57-1 | 99.9% | C30H35N7O4S | 500 MG
SDP
Catalog No. 5000357723
Encompass_Preferred
Imatinib Mesylate is an orally active tyrosine kinase inhibitor that targets c-Kit, Bcr-Abl, and PDGFR with an IC50 of approximately 100 nM. This compound is intended for research use only.
  • Inhibits c-Kit autophosphorylation, MAPK activation, and Akt activation.
  • Effective against Bcr-Abl, Kit, and PDGFR.
  • Functions as a multi-target inhibitor of v-Abl, c-Kit, Bcr/Abl, Tel/Abl, and PDGFβ receptor.
  • Does not inhibit Src family kinases, c-Fms, Flt3, or EGFR.
  • Inhibits tyrosine phosphorylation and cell growth in various cell lines.
  • Exhibits antileukemic activity in chronic myelogenous leukemia (CML) and Philadelphia-positive (Ph+) acute lymphoblastic leukemia (ALL).
Catalog No. 50-003-57723 Supplier Medchemexpress LLC Supplier No. HY50946500MG
Imatinib Mesylate is an orally active tyrosine kinase inhibitor that targets c-Kit, Bcr-Abl, and PDGFR with an IC50 of approximately 100 nM. This compound is intended for research use only.
  • Inhibits c-Kit autophosphorylation, MAPK activation, and Akt activation.
  • Effective against Bcr-Abl, Kit, and PDGFR.
  • Functions as a multi-target inhibitor of v-Abl, c-Kit, Bcr/Abl, Tel/Abl, and PDGFβ receptor.
  • Does not inhibit Src family kinases, c-Fms, Flt3, or EGFR.
  • Inhibits tyrosine phosphorylation and cell growth in various cell lines.
  • Exhibits antileukemic activity in chronic myelogenous leukemia (CML) and Philadelphia-positive (Ph+) acute lymphoblastic leukemia (ALL).