Learn More
Enzo Life Sciences Imatinib. mesylate (25mg). CAS: 220127-57-1
SDP
Supplier:  Enzo Life Sciences ALX270492M025
Encompass_Preferred
Tyrosine kinase inhibitor. Highly specific for BCR-ABL, the enzyme associated with chronic myelogenous leukemia (CML) and certain forms of acute lymphoblastic leukemia (ALL). Also shown to inhibit the transmembrane receptor kit and platelet-derived growth factor (PDGF) receptors. Alternative name: STI-571, CGP-57148B. Purity: ≥98% (HPLC). Solubility: Soluble in water (200mg/ml) or DMSO (100mg/ml) or DMF (10mg/ml) or PBS (2mg/ml). Sparingly soluble in ethanol (0.2mg/ml). Storing aqueous solutions for more than one day is not recommended. Long Term Storage: +4°C.
Catalog No. 50-200-8618