Learn More
AdipoGen Imatinib . mesylate (25 mg)
SDP
Supplier:  AdipoGen AGCR13725M025
Encompass_Preferred
Chemical. CAS: 220127-57-1. Formula: C29H31N7O . CH4SO3. MW: 493.6 . 96.1. Imatinib mesylate is a multitarget tyrosine kinase inhibitor with antineoplastic activity. It is a potent and selective inhibitor of the kinases Bcr-Abl (IC50 = 38 nM), PDGFR and c-Kit, which are encoded by the bcr-abl oncogene, as well as receptor tyrosine kinases encoded by c-kit and platelet-derived growth factor receptor (PDGFR) oncogenes, which is useful in vitro and in vivo. Imatinib inhibits ligand-stimulated autophosphorylation of PDGFR and c-Kit and the proliferation of Bcr-Abl-dependent R10(–) cells and HMC-1 cells expressing constitutively active c-Kit in a concentration-dependent manner. Imatinib inhibits replication of SARS-CoV and MERS-CoV in vitro by a novel mechanism of blocking coronavirus virion fusion with the endosomal membrane. Abl is also a potential drug target of SARS-CoV-2 and can possible inhibit the spread of COVID-19.
Catalog No. 50-205-4130