Learn More
Selleck Chemical LLC Imatinib Mesylate S1026-1g
SDP
Catalog No. 503111247
Encompass_Preferred
Imatinib Mesylate is an orally bioavailability mesylate salt of Imatinib which is a multi-target inhibitor of v-Abl c-Kit and PDGFR with IC50 of 0 6 M 0 1 M and 0 1 M in cell-free or cell-based assays respectively Imatinib Mesylate (STI571) induces autophagy
Catalog No. 50-311-1247 Supplier Selleck Chemical LLC Supplier No. S10261G
Imatinib Mesylate is an orally bioavailability mesylate salt of Imatinib which is a multi-target inhibitor of v-Abl c-Kit and PDGFR with IC50 of 0 6 M 0 1 M and 0 1 M in cell-free or cell-based assays respectively Imatinib Mesylate (STI571) induces autophagy