Learn More
Apexbio Technology LLC Imatinib Mesylate (STI571), 100mg. Cas: 220127-57-1 MFCD: MFCD04307699. Abl/c-kit/PDGFR inhibitor
SDP
Supplier:  Apexbio Technology LLC A1805100
Encompass_Preferred
Imatinib mesylate is a tyrosine kinase inhibitor IC50 value of 100 nM, 100 nM, 600 nM for v-Abl, c-kit, PDGFR, respectively [1].Tyrosine kinase is an enzyme which is a subclass of protein kinase and plays an important role in transferring a phosphate group from ATP to a protein in cells. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4710