Learn More
Selleck Chemical LLC Imatinib (STI571) Mesylate 10mM/1mL 220127-57-1 Gleevec, Glivec, CGP-57148B
SDP
Catalog No. 501360866
Encompass_Preferred
Imatinib (STI571, CGP057148B, Gleevec) Mesylate is an orally bioavailability mesylate salt of Imatinib, which is a multi-target inhibitor of v-Abl, c-Kit and PDGFR with IC50 of 0.6 ?M, 0.1 ?M and 0.1 ?M in cell-free or cell-based assays, respectively. Imatinib Mesylate (STI571) induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0866 Supplier Selleck Chemical LLC Supplier No. S102610MM/1ML
Imatinib (STI571, CGP057148B, Gleevec) Mesylate is an orally bioavailability mesylate salt of Imatinib, which is a multi-target inhibitor of v-Abl, c-Kit and PDGFR with IC50 of 0.6 ?M, 0.1 ?M and 0.1 ?M in cell-free or cell-based assays, respectively. Imatinib Mesylate (STI571) induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.