Learn More
Abcam Imatinib mesylate, Tyrosine kinase inhibitor, 100MG
Catalog No. 502706353
Encompass_Preferred
MW 589.7 Da, Purity >98%. Potent, selective inhibitor of tyrosine kinases v-Abl (IC₅₀ = 38 nM), PDGFR and c-kit. Potent inhibitor of the Bcr-Abl tyrosine kinase created by the Philadelphia chromosome abnormality found in CML. Decreases proliferation and enhances apoptosis in leukemias CML and ALL.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6353 Supplier Abcam Supplier No. AB142070100MG
MW 589.7 Da, Purity >98%. Potent, selective inhibitor of tyrosine kinases v-Abl (IC₅₀ = 38 nM), PDGFR and c-kit. Potent inhibitor of the Bcr-Abl tyrosine kinase created by the Philadelphia chromosome abnormality found in CML. Decreases proliferation and enhances apoptosis in leukemias CML and ALL.
The product is subject to the following: Abcam Restricted Use Statement