Learn More
Amsbio LLC iMatrix-511 Recombinant Laminin E8 Fragments (Laminin matrix) (2 tubes x 175μg)
Catalog No. 502041395
Encompass
iMatrix-511 is an innovative cell culture matrix compatible with a wide variety of cell types, and exceptionally well suited for pluripotent stem cells. This product is comprised of recombinant Laminin-511 E8 protein fragments which permit ES/iPS cells to be maintained in feeder-free culture conditions, enables the passaging of single cells, and provides greater adhesion than full-length Laminin, Vitronectin or Matrigel. iMatrix-511 can also be used together with AMSBIO's StemFit cell culture media and CELLBANKER cryopreservation series as part of an expanded Stem Cell Synergy solution for ES/iPS cell cultures. MAIN FEATURES: Ideal for feeder-free cell culture | Provides superior adhesion of human ES and iPS cells | Enables the passaging of single cells | Compatible across a wide variety of cell types | Makes it easy to achieve extended cultures of hES/hiPS cells | Enables efficient bulk production.
Catalog No. 50-204-1395 Supplier Amsbio LLC Supplier No. AMS.892011
iMatrix-511 is an innovative cell culture matrix compatible with a wide variety of cell types, and exceptionally well suited for pluripotent stem cells. This product is comprised of recombinant Laminin-511 E8 protein fragments which permit ES/iPS cells to be maintained in feeder-free culture conditions, enables the passaging of single cells, and provides greater adhesion than full-length Laminin, Vitronectin or Matrigel. iMatrix-511 can also be used together with AMSBIO's StemFit cell culture media and CELLBANKER cryopreservation series as part of an expanded Stem Cell Synergy solution for ES/iPS cell cultures. MAIN FEATURES: Ideal for feeder-free cell culture | Provides superior adhesion of human ES and iPS cells | Enables the passaging of single cells | Compatible across a wide variety of cell types | Makes it easy to achieve extended cultures of hES/hiPS cells | Enables efficient bulk production.