Learn More
Iwai North America iMatrix-511 SILK (Transgenic Silkworm Cocoon)(Size 1050ug)
Catalog No. NC1572446
Encompass_Preferred
iMatrix-511 is a recombinant Laminin- 511 E8 fragment expressed by transgenic silkworm cocoon. Laminin-511 E8 fragments allow ES/iPS cells and MSCs to be maintained in xeno-free culture conditions. It also enables the passaging of single cells and provides greater adhesion than full-length Laminin, Vitronectin, or Matrigel. 2) This is an in stock item and lead time should be similar to what you have for N-892012. Currently it indicates ships in 6 business days but it close to 2 business days.
Catalog No. NC1572446 Supplier Iwai North America Supplier No. N892021
iMatrix-511 is a recombinant Laminin- 511 E8 fragment expressed by transgenic silkworm cocoon. Laminin-511 E8 fragments allow ES/iPS cells and MSCs to be maintained in xeno-free culture conditions. It also enables the passaging of single cells and provides greater adhesion than full-length Laminin, Vitronectin, or Matrigel. 2) This is an in stock item and lead time should be similar to what you have for N-892012. Currently it indicates ships in 6 business days but it close to 2 business days.