Learn More
Amsbio LLC Recombinant Laminin iMatrix-511 silk (6 tubes x 175μg)
Catalog No. 502041396
Encompass
iMatrix-511 is an innovative cell culture matrix comprised of recombinant Laminin-511 E8 protein fragments expressed in silkworms which permit ES/iPS cells to be maintained in feeder-free culture conditions. It also enables the passaging of single cells, & provides greater adhesion than full-length Laminin, Vitronectin or Matrigel. iMatrix-511 can also be used together with AMSBIO's StemFit cell culture media and CELLBANKER cryopreservation series as part of an expanded Stem Cell Synergy solution for ES/iPS cell cultures.
Catalog No. 50-204-1396 Supplier Amsbio LLC Supplier No. AMS.892021
iMatrix-511 is an innovative cell culture matrix comprised of recombinant Laminin-511 E8 protein fragments expressed in silkworms which permit ES/iPS cells to be maintained in feeder-free culture conditions. It also enables the passaging of single cells, & provides greater adhesion than full-length Laminin, Vitronectin or Matrigel. iMatrix-511 can also be used together with AMSBIO's StemFit cell culture media and CELLBANKER cryopreservation series as part of an expanded Stem Cell Synergy solution for ES/iPS cell cultures.