Learn More
Foxx Life Sciences Imaweld Heat Sealable and Weldable TPE Tube 9.5mm (3/8") ID X 15.87mm (5/8") OD, 50 FT
GREENER_CHOICE
Catalog No. 502061523
Encompass_Preferred
Imaweld is thermoplastic elastomer opaque tubing designed for fluid transfer in pharmaceutical and biotech applications. Imaweld is specially formulated which meets requirement of pharmaceutical industries with superiority compare to PVC and Silicone.• Free of any toxic ingredients. successfully meets toxicity free requirements.• Manufactured and Packaged in Clean Room Class of ISO 9001, ISO 14001 and OHSAS 18001 certified facility• Heat sealable and weldable to facilitate sterile connection• Working Temperature Range -40°C to +135°C• USP Class VI Certification• TSE/BSE CertificationAll standard dimensions are in mm and Inch dimensions are for reference only.
Catalog No. 50-206-1523 Supplier Foxx Life Sciences Supplier No. WEL00950159050
Imaweld is thermoplastic elastomer opaque tubing designed for fluid transfer in pharmaceutical and biotech applications. Imaweld is specially formulated which meets requirement of pharmaceutical industries with superiority compare to PVC and Silicone.• Free of any toxic ingredients. successfully meets toxicity free requirements.• Manufactured and Packaged in Clean Room Class of ISO 9001, ISO 14001 and OHSAS 18001 certified facility• Heat sealable and weldable to facilitate sterile connection• Working Temperature Range -40°C to +135°C• USP Class VI Certification• TSE/BSE CertificationAll standard dimensions are in mm and Inch dimensions are for reference only.