Learn More
Lkt Laboratories Imazalil, ≥95%, 10GR
SDP
Supplier:  Lkt Laboratories I507210G
Encompass_Preferred
Triazole; 14-α demethylase and aromatase inhibitor, AR antagonist. CAS: 35554-44-0. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4935