Learn More
Cayman Chemical Imazalil-d5 (sulfate), Purity: ≥99% deuterated forms (d1-d5), CAS: 1398065-92-3, 5 mg
SDP
Catalog No. 502916877
Encompass_Preferred
An internal standard for the quantification of imazalil by GC- or LC-MS
Catalog No. 50-291-6877 Supplier Cayman Chemical Supplier No. 392615 MG
An internal standard for the quantification of imazalil by GC- or LC-MS