Learn More
Medchemexpress LLC Imazalil sulfate | 58594-72-2 | 99.6% | 5 G
SDP
Catalog No. 5000413722
Encompass_Preferred
Imazalil (Enilconazole) sulfate is a fungicide. Imazalil sulfate has oral activity and strongly activates mPXR but not mCAR in mouse liver. It is commonly used to protect various agricultural crops against fungal attack. Imazalil sulfate induces developmental abnormalities, gut microbiota dysbiosis, and hepatic metabolism disorder.
  • Fungicide
  • Oral activity
  • Strongly activates mPXR but not mCAR in mouse liver
  • Commonly used to protect various agricultural crops against fungal attack
  • Induces developmental abnormalities, gut microbiota dysbiosis, and hepatic metabolism disorder
Catalog No. 50-004-13722 Supplier Medchemexpress LLC Supplier No. HYB1134A5G
Imazalil (Enilconazole) sulfate is a fungicide. Imazalil sulfate has oral activity and strongly activates mPXR but not mCAR in mouse liver. It is commonly used to protect various agricultural crops against fungal attack. Imazalil sulfate induces developmental abnormalities, gut microbiota dysbiosis, and hepatic metabolism disorder.
  • Fungicide
  • Oral activity
  • Strongly activates mPXR but not mCAR in mouse liver
  • Commonly used to protect various agricultural crops against fungal attack
  • Induces developmental abnormalities, gut microbiota dysbiosis, and hepatic metabolism disorder