Learn More
TARGETMOL CHEMICALS INC Imazodan hydrochloride 1MG
SDP
Catalog No. 502879423
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Imazodan hydrochloride (CI-914 HCl) is a potent and selective type III phosphodiesterase inhibitor used in the treatment of chronic congestive heart failure. Purity 99.84%
Catalog No. 50-287-9423 Supplier TARGETMOL CHEMICALS INC Supplier No. T275961MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Imazodan hydrochloride (CI-914 HCl) is a potent and selective type III phosphodiesterase inhibitor used in the treatment of chronic congestive heart failure. Purity 99.84%