Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Imb-301 | 64009-84-3 | 99.1% | 10 MG
SDP

Catalog No. 5000231126
Encompass_Preferred

IMB-301 is a small-molecule inhibitor of HIV-1 replication that binds human APOBEC3G (hA3G), disrupts the hA3G-Vif interaction, and inhibits Vif-mediated degradation of hA3G; it has reported activity inhibiting HIV-1 replication in H9 cells (IC50 = 8.63 μM).

  • Small-molecule HIV-1 replication inhibitor.
  • Binds hA3G and disrupts hA3G-Vif interaction.
  • Reported IC50 of 8.63 μM in H9 cells.
  • Molecular weight 379.26 g/mol.
  • High reported purity (99.08%).
  • Storage: pure form at -20°C; in solvent at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-31126 Supplier Medchemexpress LLC Supplier No. HY12215610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IMB-301 is a small-molecule inhibitor of HIV-1 replication that binds human APOBEC3G (hA3G), disrupts the hA3G-Vif interaction, and inhibits Vif-mediated degradation of hA3G; it has reported activity inhibiting HIV-1 replication in H9 cells (IC50 = 8.63 μM).

  • Small-molecule HIV-1 replication inhibitor.
  • Binds hA3G and disrupts hA3G-Vif interaction.
  • Reported IC50 of 8.63 μM in H9 cells.
  • Molecular weight 379.26 g/mol.
  • High reported purity (99.08%).
  • Storage: pure form at -20°C; in solvent at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.