Learn More
IMCS Rapid Hydrolysis Buffer, 100mL
SDP
Catalog No. NC1689174
Encompass
Rapid Hydrolysis Buffer is filter sterilized and designed to maintain optimal pH for efficient hydrolysis of IMCSzyme E1F, E1FG and 3S products.
Catalog No. NC1689174 Supplier IMCS Supplier No. 04EZRHBA100
Rapid Hydrolysis Buffer is filter sterilized and designed to maintain optimal pH for efficient hydrolysis of IMCSzyme E1F, E1FG and 3S products.