Learn More
Medchemexpress LLC HY-10172 50mg , IMD-0354 CAS:978-62-1 Purity:98%
SDP
Catalog No. 501493710
Encompass_Preferred
Medchemexpress, HY-10172 50mg IMD-0354 CAS:978-62-1 Formula:C15H8ClF6NO2 IKKβ IC50: 1.2±0.3 μM (NF-κB) Purity:98% IMD-0354 is a selective IKKβ inhibitor which inhibits NF-κB activity. IMD0354 inhibits TNF-α induced NF-κB transcription activity with an IC 50 of 1.2±0.3 uM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3710 Supplier Medchemexpress LLC Supplier No. HY1017250MG
Medchemexpress, HY-10172 50mg IMD-0354 CAS:978-62-1 Formula:C15H8ClF6NO2 IKKβ IC50: 1.2±0.3 μM (NF-κB) Purity:98% IMD-0354 is a selective IKKβ inhibitor which inhibits NF-κB activity. IMD0354 inhibits TNF-α induced NF-κB transcription activity with an IC 50 of 1.2±0.3 uM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.