Learn More
Selleck Chemical LLC IMD 0354 5mg 978-62-1
SDP
Supplier:  Selleck Chemical LLC S28645MG
Encompass_Preferred
IMD-0354 is an IKK? inhibitor and blocks I?B? phosphorylation in NF-?B pathway. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3094