Learn More
TARGETMOL CHEMICALS INC IMDK 5MG
SDP
Catalog No. 502576202
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. iMDK quarterhydrate is a potent PI3K inhibitor that inhibits the growth factor MDK (also known as midkine or MK). iMDK quarterhydrate synergistically inhibits non-small cell lung cancer (NSCLC) with MEK inhibitors without harming normal cells and mice. purity: 99%
Catalog No. 50-257-6202 Supplier TARGETMOL CHEMICALS INC Supplier No. T94605MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. iMDK quarterhydrate is a potent PI3K inhibitor that inhibits the growth factor MDK (also known as midkine or MK). iMDK quarterhydrate synergistically inhibits non-small cell lung cancer (NSCLC) with MEK inhibitors without harming normal cells and mice. purity: 99%