Learn More
STEMCELL Technologies Iscove's Modified Dulbecco's Medium, Size: 500 mL
SDP
Catalog No. NC9824854
Encompass
Iscove's Modified Dulbecco's Medium (IMDM) with 25 mM HEPES is recommended for a wide variety of cell culture applications. Selection of suitable nutrient medium is dependent on the type of cell, conditions of culture, and degree of chemical definition required for the cell culture application.
Catalog No. NC9824854 Supplier STEMCELL Technologies Supplier No. 36150
Iscove's Modified Dulbecco's Medium (IMDM) with 25 mM HEPES is recommended for a wide variety of cell culture applications. Selection of suitable nutrient medium is dependent on the type of cell, conditions of culture, and degree of chemical definition required for the cell culture application.