Learn More
ATHENA ES IMDM, 500 ml
Catalog No. 5000158067
Encompass_Preferred
Iscove’s Modified Dulbecco’s Medium (IMDM) is a standard serum-free medium. It is supplemented with bovine pituitary extract and epidermal growth factor and contains L-glutamine and Hepes but no antibiotics.
Catalog No. 50-001-58067 Supplier ATHENA ES Supplier No. 0411
Iscove’s Modified Dulbecco’s Medium (IMDM) is a standard serum-free medium. It is supplemented with bovine pituitary extract and epidermal growth factor and contains L-glutamine and Hepes but no antibiotics.