Learn More
STEMCELL Technologies Iscove's MDM with 2% FBS, Size: 100 mL
SDP
Catalog No. NC9968227
Encompass
Iscove's Modified Dulbecco's Medium (MDM) with 2% Fetal Bovine Serum (FBS) is recommended for preparing and washing human cord blood, peripheral blood, or bone marrow samples for colony-forming unit (CFU) assays.
Catalog No. NC9968227 Supplier STEMCELL Technologies Supplier No. 07700
Iscove's Modified Dulbecco's Medium (MDM) with 2% Fetal Bovine Serum (FBS) is recommended for preparing and washing human cord blood, peripheral blood, or bone marrow samples for colony-forming unit (CFU) assays.