Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech IMDM Cell Culture Medium, w: L-Glutamine, w: 25 mM HEPES, w: 3.024 g/L NaHCO3 (500 ml)
SDP
Catalog No. 502388487
Encompass_Preferred
IMDM, WITH: L-Glutamine, WITH: 25 mM HEPES, WITH: 3.024 g/L NaHCO3. QUANTITY: 500 ml. MANUFACTURED BY: Pan-Biotech (Germany). STORAGE: 2 to 8 degrees Celsius. DESCRIPTION: Iscove's Modified Dulbecco's Medium (IMDM) is a modification of Dulbecco's Modified Eagle Medium (DMEM) that includes selenium, additional amino acids and vitamins, sodium pyruvate, and HEPES buffer. Furthermore, in IMDM, ferric nitrate (iron) is replaced with potassium nitrate. IMDM is well suited for rapidly proliferating cultures of high cell density. When supplemented with albumin, transferrin, and soy lipids it can be excellently applied for culturing lymphocytes, marrow cells, or hybridoma cells.
Catalog No. 50-238-8487 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0420150
IMDM, WITH: L-Glutamine, WITH: 25 mM HEPES, WITH: 3.024 g/L NaHCO3. QUANTITY: 500 ml. MANUFACTURED BY: Pan-Biotech (Germany). STORAGE: 2 to 8 degrees Celsius. DESCRIPTION: Iscove's Modified Dulbecco's Medium (IMDM) is a modification of Dulbecco's Modified Eagle Medium (DMEM) that includes selenium, additional amino acids and vitamins, sodium pyruvate, and HEPES buffer. Furthermore, in IMDM, ferric nitrate (iron) is replaced with potassium nitrate. IMDM is well suited for rapidly proliferating cultures of high cell density. When supplemented with albumin, transferrin, and soy lipids it can be excellently applied for culturing lymphocytes, marrow cells, or hybridoma cells.