Learn More
ILEX LIFE SCIENCES LLC IMDM P04-20250 6 X 500 ML
SDP
Catalog No. 502733825
Encompass_Preferred
PAN-Biotech IMDM Cell Culture Medium, w/o: L-Glutamine, w: 3.024 g/L NaHCO3 (6 x 500 ml). BICARBONATE: With 3.024 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: Without glutamine. PHENOL RED: With Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: With sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Iscove's Modified Dulbecco's Medium (IMDM) is a modification of Dulbecco's Modified Eagle Medium (DMEM) that includes selenium, additional amino acids and vitamins, and sodium pyruvate. Furthermore, in IMDM, ferric nitrate (iron) is replaced with potassium nitrate. IMDM is well suited for rapidly proliferating cultures of high cell density. When supplemented with albumin, transferrin, and soy lipids it can be excellently applied for culturing lymphocytes, marrow cells, or hybridoma cells.
Catalog No. 50-273-3825 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0420250X6
Add to Cart
Add to Cart
PAN-Biotech IMDM Cell Culture Medium, w/o: L-Glutamine, w: 3.024 g/L NaHCO3 (6 x 500 ml). BICARBONATE: With 3.024 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: Without glutamine. PHENOL RED: With Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: With sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Iscove's Modified Dulbecco's Medium (IMDM) is a modification of Dulbecco's Modified Eagle Medium (DMEM) that includes selenium, additional amino acids and vitamins, and sodium pyruvate. Furthermore, in IMDM, ferric nitrate (iron) is replaced with potassium nitrate. IMDM is well suited for rapidly proliferating cultures of high cell density. When supplemented with albumin, transferrin, and soy lipids it can be excellently applied for culturing lymphocytes, marrow cells, or hybridoma cells.