Learn More
ILEX LIFE SCIENCES LLC IMDM P04-20450 6 X 500 ML
SDP
Catalog No. 502733819
Encompass_Preferred
PAN-Biotech IMDM Cell Culture Medium, w: stable Glutamine, w: 25 mM HEPES, w: 3.024 g/L NaHCO3 (6 x 500 ml). BICARBONATE: With 3.024 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: With Stable Glutamine. PHENOL RED: With Phenol Red. HEPES: With 25 mM HEPES. SODIUM PYRUVATE: With sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Iscove's Modified Dulbecco's Medium (IMDM) is a modification of Dulbecco's Modified Eagle Medium (DMEM) that includes selenium, additional amino acids and vitamins, and sodium pyruvate. Furthermore, in IMDM, ferric nitrate (iron) is replaced with potassium nitrate. IMDM is well suited for rapidly proliferating cultures of high cell density. When supplemented with albumin, transferrin, and soy lipids it can be excellently applied for culturing lymphocytes, marrow cells, or hybridoma cells.
Catalog No. 50-273-3819 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0420450X6
Add to Cart
Add to Cart
PAN-Biotech IMDM Cell Culture Medium, w: stable Glutamine, w: 25 mM HEPES, w: 3.024 g/L NaHCO3 (6 x 500 ml). BICARBONATE: With 3.024 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: With Stable Glutamine. PHENOL RED: With Phenol Red. HEPES: With 25 mM HEPES. SODIUM PYRUVATE: With sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Iscove's Modified Dulbecco's Medium (IMDM) is a modification of Dulbecco's Modified Eagle Medium (DMEM) that includes selenium, additional amino acids and vitamins, and sodium pyruvate. Furthermore, in IMDM, ferric nitrate (iron) is replaced with potassium nitrate. IMDM is well suited for rapidly proliferating cultures of high cell density. When supplemented with albumin, transferrin, and soy lipids it can be excellently applied for culturing lymphocytes, marrow cells, or hybridoma cells.