Learn More
IMEB, Inc Hydrochloric Acid/Formic Acid Decalcification Solution, 1 Gallon
SDP
Catalog No. NC9875165
Encompass_Preferred
Hydrochloric Acid/Formic Acid Decalcification Solution: a combination to illustrate the best decalcifying agent by Lillie. Formic acid permits satisfactory nuclear and marrow cell staining, while Hydrochloric Acid rapidly decalcifies even compact bone in 12 to 24 hours. As with any acid solution, swelling of tissue is characteristic after extended periods. Reagent alcohol may be added to solution to remedy the swelling action.
Catalog No. NC9875165 Supplier IMEB, Inc Supplier No. HA100
Hydrochloric Acid/Formic Acid Decalcification Solution: a combination to illustrate the best decalcifying agent by Lillie. Formic acid permits satisfactory nuclear and marrow cell staining, while Hydrochloric Acid rapidly decalcifies even compact bone in 12 to 24 hours. As with any acid solution, swelling of tissue is characteristic after extended periods. Reagent alcohol may be added to solution to remedy the swelling action.