Learn More
TARGETMOL CHEMICALS INC IMEGLIMIN HYDROCHLORIDE 10MG
SDP
Catalog No. 502576312
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Imeglimin hydrochloride (EMD 387008 hydrochloride) prevents human endothelial cell death by inhibiting mitochondrial permeability transition without inhibiting mitochondrial respiration. purity: 99%
Catalog No. 50-257-6312 Supplier TARGETMOL CHEMICALS INC Supplier No. T748610MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Imeglimin hydrochloride (EMD 387008 hydrochloride) prevents human endothelial cell death by inhibiting mitochondrial permeability transition without inhibiting mitochondrial respiration. purity: 99%