Learn More
Apexbio Technology LLC Imeglimin hydrochloride 775351-61-6 100mg
SDP
Catalog No. 5000094037
Encompass_Preferred
Imeglimin hydrochloride (CAS 775351-61-6) is a small molecule investigated as an oral agent for type 2 diabetes It acts by modulating mitochondrial function across hepatic muscular and pancreatic tissues In hepatocytes imeglimin redirects substrate flow towards complex II inhibits complex I restores complex III activity and enhances fatty acid oxidation which is supported by increased 3-hydroxyacyl-CoA dehydrogenase activity and acylcarnitine levels The compound also reduces reactive oxygen species and elevates mitochondrial DNA content In high-fat high-sucrose diet mouse models imeglimin improves glucose tolerance insulin sensitivity and mitochondrial function while normalizing metabolic parameters Currently imeglimin is under clinical evaluation both as monotherapy and in combination with other antidiabetic agents
Catalog No. 50-000-94037 Supplier Apexbio Technology LLC Supplier No. B1067100
Imeglimin hydrochloride (CAS 775351-61-6) is a small molecule investigated as an oral agent for type 2 diabetes It acts by modulating mitochondrial function across hepatic muscular and pancreatic tissues In hepatocytes imeglimin redirects substrate flow towards complex II inhibits complex I restores complex III activity and enhances fatty acid oxidation which is supported by increased 3-hydroxyacyl-CoA dehydrogenase activity and acylcarnitine levels The compound also reduces reactive oxygen species and elevates mitochondrial DNA content In high-fat high-sucrose diet mouse models imeglimin improves glucose tolerance insulin sensitivity and mitochondrial function while normalizing metabolic parameters Currently imeglimin is under clinical evaluation both as monotherapy and in combination with other antidiabetic agents