Learn More
Medchemexpress LLC Imeglimin (hydrochloride) | 775351-61-6 | 99.3% | 191.66 | 1 ML
SDP
Catalog No. 5000361592
Encompass_Preferred
Imeglimin hydrochloride is an oral glucose-lowering agent known for its ability to reduce reactive oxygen species (ROS) production, increase mitochondrial DNA, and improve mitochondrial function. This specific product is provided as a 10 mM solution in 1 mL of DMSO, suitable for various research applications. It is intended for research use only and has not been validated for medical applications.
  • Reduces reactive oxygen species production
  • Increases mitochondrial DNA
  • Improves mitochondrial function
  • Provided as a 10 mM solution in 1 mL DMSO
  • Intended for research use only
Catalog No. 50-003-61592 Supplier Medchemexpress LLC Supplier No. HY14771A10MM/1ML
Imeglimin hydrochloride is an oral glucose-lowering agent known for its ability to reduce reactive oxygen species (ROS) production, increase mitochondrial DNA, and improve mitochondrial function. This specific product is provided as a 10 mM solution in 1 mL of DMSO, suitable for various research applications. It is intended for research use only and has not been validated for medical applications.
  • Reduces reactive oxygen species production
  • Increases mitochondrial DNA
  • Improves mitochondrial function
  • Provided as a 10 mM solution in 1 mL DMSO
  • Intended for research use only