Learn More
Medchemexpress LLC Imeglimin (hydrochloride) | 775351-61-6 | 99.3% | 191.66 | 25 MG
SDP
Catalog No. 5000362916
Encompass_Preferred
Imeglimin hydrochloride (EMD 387008) is an oral glucose-lowering agent that reduces reactive oxygen species (ROS) production, increases mitochondrial DNA, and improves mitochondrial function. This compound appears as a white to off-white solid.
  • Oral glucose-lowering agent
  • Reduces reactive oxygen species (ROS) production
  • Increases mitochondrial DNA
  • Improves mitochondrial function
  • Soluble in H2O (≥ 50 mg/mL)
  • Soluble in DMSO (25 mg/mL)
Catalog No. 50-003-62916 Supplier Medchemexpress LLC Supplier No. 25MGHY14771A
Imeglimin hydrochloride (EMD 387008) is an oral glucose-lowering agent that reduces reactive oxygen species (ROS) production, increases mitochondrial DNA, and improves mitochondrial function. This compound appears as a white to off-white solid.
  • Oral glucose-lowering agent
  • Reduces reactive oxygen species (ROS) production
  • Increases mitochondrial DNA
  • Improves mitochondrial function
  • Soluble in H2O (≥ 50 mg/mL)
  • Soluble in DMSO (25 mg/mL)