Learn More
Medchemexpress LLC Imeglimin (hydrochloride) | 775351-61-6 | 99.3% | 191.66 | 50 MG
SDP
Catalog No. 5000362824
Encompass_Preferred
Imeglimin hydrochloride (EMD 387008) is an oral glucose-lowering agent. It also reduces reactive oxygen species (ROS) production, increases mitochondrial DNA, and improves mitochondrial function.
  • Oral glucose-lowering agent
  • Reduces reactive oxygen species production
  • Increases mitochondrial DNA
  • Improves mitochondrial function
Catalog No. 50-003-62824 Supplier Medchemexpress LLC Supplier No. HY14771A50MG
Imeglimin hydrochloride (EMD 387008) is an oral glucose-lowering agent. It also reduces reactive oxygen species (ROS) production, increases mitochondrial DNA, and improves mitochondrial function.
  • Oral glucose-lowering agent
  • Reduces reactive oxygen species production
  • Increases mitochondrial DNA
  • Improves mitochondrial function