Learn More
Medchemexpress LLC Imeglimin hydrochloride | 775351-61-6 | MFCD28167741 | 100.0% | 191.66 g/mol | C6H14ClN5 | 10 MG
SDP
Catalog No. 5000287460
Encompass_Preferred
Imeglimin hydrochloride is the hydrochloride salt of imeglimin, an oral glucose-lowering small molecule that modulates mitochondrial metabolism and improves insulin sensitivity. It is supplied as a research-grade reagent for in vitro and in vivo studies of glucose homeostasis and mitochondrial function.
  • High purity suitable for research applications.
  • Molecular weight 191.66 g/mol and chemical formula C6H14ClN5.
  • Provided as a solid in a 10 MG pack.
  • Used to study insulin sensitivity and mitochondrial metabolism.
  • Includes a product datasheet with analytical information.
Catalog No. 50-002-87460 Supplier Medchemexpress LLC Supplier No. HY14771A10MG
Imeglimin hydrochloride is the hydrochloride salt of imeglimin, an oral glucose-lowering small molecule that modulates mitochondrial metabolism and improves insulin sensitivity. It is supplied as a research-grade reagent for in vitro and in vivo studies of glucose homeostasis and mitochondrial function.
  • High purity suitable for research applications.
  • Molecular weight 191.66 g/mol and chemical formula C6H14ClN5.
  • Provided as a solid in a 10 MG pack.
  • Used to study insulin sensitivity and mitochondrial metabolism.
  • Includes a product datasheet with analytical information.