Learn More
Medchemexpress LLC HY-14953 5mg Medchemexpress, Imepitoin CAS:188116-07-6 Purity:>98%
SDP
Catalog No. 502081666
Encompass_Preferred
Medchemexpress, HY-14953 5mg Imepitoin CAS:188116-07-6 Imepitoin (AWD 131-138) is a new low-affinity partial benzodiazepine receptor agonist with potent anticonvulsant and anxiolytic properties in rodent models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1666 Supplier Medchemexpress LLC Supplier No. HY149535MG
Medchemexpress, HY-14953 5mg Imepitoin CAS:188116-07-6 Imepitoin (AWD 131-138) is a new low-affinity partial benzodiazepine receptor agonist with potent anticonvulsant and anxiolytic properties in rodent models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.