Learn More
Cayman Chemical ImethrIdInhydrobromIde 10mg
SDP
Catalog No. 502990204
Encompass_Preferred
A potent histamine H3 receptor agonist (Ki = 0.85 nM); selective for histamine H3 over histamine H4 receptors (Ki = 245 nM); inhibits cAMP-stimulated β-galactosidase transcription in SK-N-MC cells expressing human H3 receptors (EC50 = 0.18 nM); induces a decrease of the electrically induced twitch contraction in isolated guinea pig ileum; inhibits hydrochloric acid-induced formation of gastric lesions in rats at 30 mg/kg, s.c.
Catalog No. 50-299-0204 Supplier Cayman Chemical Supplier No. 2950210 MG
A potent histamine H3 receptor agonist (Ki = 0.85 nM); selective for histamine H3 over histamine H4 receptors (Ki = 245 nM); inhibits cAMP-stimulated β-galactosidase transcription in SK-N-MC cells expressing human H3 receptors (EC50 = 0.18 nM); induces a decrease of the electrically induced twitch contraction in isolated guinea pig ileum; inhibits hydrochloric acid-induced formation of gastric lesions in rats at 30 mg/kg, s.c.