Learn More
Apexbio Technology LLC Imetit dihydrobromide 32385-58-3 5mg
SDP
Catalog No. 5000092063
Encompass_Preferred
Imetit dihydrobromide (CAS 32385-58-3) is a potent high-affinity agonist of histamine H3 and H4 receptors demonstrating Ki values of 0 3 nM and 2 7 nM respectively Functionally it replicates histaminergic effects by inducing morphological changes in eosinophils with an EC50 of 25 nM Owing to its selective receptor activity Imetit dihydrobromide is widely employed in studies investigating allergic inflammatory processes including asthma as well as in research focused on central nervous system physiology and histamine-related pathways
Catalog No. 50-000-92063 Supplier Apexbio Technology LLC Supplier No. B64245
Imetit dihydrobromide (CAS 32385-58-3) is a potent high-affinity agonist of histamine H3 and H4 receptors demonstrating Ki values of 0 3 nM and 2 7 nM respectively Functionally it replicates histaminergic effects by inducing morphological changes in eosinophils with an EC50 of 25 nM Owing to its selective receptor activity Imetit dihydrobromide is widely employed in studies investigating allergic inflammatory processes including asthma as well as in research focused on central nervous system physiology and histamine-related pathways