Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Imetra Diamond Scribes

Catalog No. 08675 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08-675 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-675

missing translation for 'validMainframeMsg'

Diamond tip writing instrument provides comfortable grip for etching glass and other surfaces

Sturdy etching instrument designed to look and feel like a pen

  • Diamond tip
  • Aluminum shaft
  • Clip
  • Barrel's shape: Hexagonal
  • Cap included

Specifications

Product Type Scribe Pen
Color Silver
Tip Material Diamond
For Use With (Application) Glass, ceramic, metal, and other.
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.