Learn More
Hach Company Cone, Imhoff Settling, 1 L, Glass
Supplier:  Hach Company 57100
Encompass
Fully graduated cones for determination of small amounts of settleable matter in sewage by Imhoff Sedimentation Method. Glass cone features reinforced tip for ruggedness, while the Polycarbonate cone features a leak-proof, removable closure for withdrawal of sediment for analysis. Support ring, support stand, and cone holders available and sold separately.
Catalog No. 50-199-4396