Learn More
Hach Company Imhoff Cone Support Stand, Three Cones, Heavy Gauge Steel
Catalog No. 501994397 Shop All Hach Company Products
Encompass
Cone support stand holds three glass or plastic cones (P/N 571-00 and P/N 2067-00, respectively).
Catalog No. 50-199-4397 Supplier Hach Company Supplier No. 57200
Cone support stand holds three glass or plastic cones (P/N 571-00 and P/N 2067-00, respectively).