Learn More
International Medical Industries (IMI) Filtered Venting Needle 20 Gauge X 1.5 Inch (Box of 100)
Catalog No. NC1868463
Encompass
Rx-Vent™ Filtered Venting Needle is used by Pharmacists and other specialists to vent evacuated containers and to maintain the necessary sterility. The non-coring needle easily penetrates all stoppers with minimum risk of damaged and the 0.2-micron hydrophobic filter helps maintain asepsis. • tainless Steel Needle • Sterile • Individual Blisters •Box of 100 • 20 Gauge x 1.5” (3.8 CM) • 0.2-micron hydrophobic filter
This item is not returnable. View return policy
Catalog No. NC1868463 Supplier International Medical Industries (IMI) Supplier No. 3120
 Restricted Item
Add to Cart
Add to Cart
Rx-Vent™ Filtered Venting Needle is used by Pharmacists and other specialists to vent evacuated containers and to maintain the necessary sterility. The non-coring needle easily penetrates all stoppers with minimum risk of damaged and the 0.2-micron hydrophobic filter helps maintain asepsis. • tainless Steel Needle • Sterile • Individual Blisters •Box of 100 • 20 Gauge x 1.5” (3.8 CM) • 0.2-micron hydrophobic filter