Learn More
FUJIFILM Biosciences Imibenconazole Standard
Catalog No. 5000450340
Encompass_Preferred
This standard-grade Imibenconazole is ideal for analytical and quality control use in pesticide residue testing Its confirmed purity and stability make it suitable for sensitive detection methods such as gas chromatography and quantitative NMR
Catalog No. 50-004-50340 Supplier FUJIFILM Biosciences Supplier No. 09504091
This standard-grade Imibenconazole is ideal for analytical and quality control use in pesticide residue testing Its confirmed purity and stability make it suitable for sensitive detection methods such as gas chromatography and quantitative NMR