Learn More
FUJIFILM Biosciences Imicyafos Standard
Catalog No. 5000450390
Encompass_Preferred
Imicyafos standard is supplied at 97% purity verified via both HPLC and quantitative NMR Recommended for use in environmental and pesticide residue testing under laboratory conditions
Catalog No. 50-004-50390 Supplier FUJIFILM Biosciences Supplier No. 09106911
Imicyafos standard is supplied at 97% purity verified via both HPLC and quantitative NMR Recommended for use in environmental and pesticide residue testing under laboratory conditions