Learn More
Apexbio Technology LLC Imidacloprid 138261-41-3 100mg
SDP
Catalog No. 5000092730
Encompass_Preferred
Imidacloprid is a small-molecule modulator targeting nicotinic acetylcholine receptors (nAChRs) in insect nervous systems It is designed to selectively bind to nAChRs thereby disrupting cholinergic neurotransmission and neuronal signaling Imidacloprid exerts its biological activity primarily through interference with normal synaptic transmission by binding to nAChRs and causing abnormal neuronal signaling Based on these pharmacological properties Imidacloprid holds research potential in investigating cholinergic neurotransmission pathways receptor-ligand interactions neuronal signaling dysfunction neurotoxicity mechanisms and insecticide resistance It is also used as a comparative or reference molecule when characterizing the pharmacological or neurotoxicological profiles of candidate neuroactive compounds and insecticidal agents
Catalog No. 50-000-92730 Supplier Apexbio Technology LLC Supplier No. C6719100
Imidacloprid is a small-molecule modulator targeting nicotinic acetylcholine receptors (nAChRs) in insect nervous systems It is designed to selectively bind to nAChRs thereby disrupting cholinergic neurotransmission and neuronal signaling Imidacloprid exerts its biological activity primarily through interference with normal synaptic transmission by binding to nAChRs and causing abnormal neuronal signaling Based on these pharmacological properties Imidacloprid holds research potential in investigating cholinergic neurotransmission pathways receptor-ligand interactions neuronal signaling dysfunction neurotoxicity mechanisms and insecticide resistance It is also used as a comparative or reference molecule when characterizing the pharmacological or neurotoxicological profiles of candidate neuroactive compounds and insecticidal agents